Oakton Replacement pH Sensor for Testr 3, 3+, 10, 20, and 30; DJ
Representative image only.
Oakton Item # EW-35624-38
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample
  • Choose from a variety of accessories for Oakton® meters
  • For pHtestr 30 purchased before Oct. 2019
  • 6 month warranty
PRODUCT OPTIONS
$104.85USD / Each
$116.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton pHTestr 3, 3+, 10, 20 & 30
  • DescriptionReplacement pH Sensor for Testr 3, 3+, 10, 20, and 30; DJ
  • Warranty6 month
6 Month Warranty
Key Features
  • Choose from a variety of accessories for Oakton® meters
  • For pHtestr 30 purchased before Oct. 2019
  • 6 month warranty
More About this Item
For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.