Environmental Express PH300 pH Sensor Cartridge, Double-Junction
Representative image only.
Environmental Express Item # EW-35660-92
Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
PRODUCT OPTIONS
$219.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypepH Sensor Cartridge
  • Range0 to 14 pH
  • Materialglass, silicone rubber, polycarbonate, polyethlene, and Polyolefin
  • Length (cm)11
  • DescriptionpH Sensor Cartridge, Double-Junction
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
More About this Item
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.