Environmental Express pH 100 Portable pH Meter with ATC Temperature Probe

Representative image only.
Environmental Express Item # EW-35613-28
Convenient, compact design with large display
  • Measured parameter and temperature shown on display at same time
  • Utilizes same electrodes as Oakton 6+ series
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • 2 year warranty

PRODUCT OPTIONS

$334.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypepH, mV, ORP, Temperature
  • Display typeLCD
  • ConnectionsBNC & Φ2.5 socket
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1 pH
  • pH Accuracy±0.01 pH
  • Buffer RecognitionAutomatic recognition of USA, NIST, and user-defined pH calibration solutions
  • Calibration1-3 points auto calibration
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution0.1 mV from –200 to 200 mV; 1 mV remaining
  • mV Accuracy±0.2% full-scale
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±0.9°F (0.5°C) ±1 digit
  • Temperature compensation0 to 100˚C (manual or automatic)
  • Temperature Coefficient2.0%/°C
  • WaterproofIP57
  • Battery3 AA (included)
  • Battery life (hr)400
  • Length (cm)9.1
  • Width (cm)19
  • Height (cm)3.3
  • Width (in)7 31/64
  • Length (in)3 37/64
  • Height (in)1 19/64
  • Stirring ControlNo
  • NIST Calibration IncludedNo
  • Probe IncludedTemperature (ATC)
  • Accessories IncludedNone
  • DescriptionOakton pH100 Portable pH Meter with Temperature Probes
  • Warranty2 year
Two year warrantyConformite Europeenne

Includes

pH 100 portable pH meter, temperature probe, and 3 AA batteries.

Key Features

  • Measured parameter and temperature shown on display at same time
  • Utilizes same electrodes as Oakton 6+ series
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • 2 year warranty

More About this Item

The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is 2.75 in. by 2.0 in. for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The pH 100 meter measures pH, mV, ORP, and temperature.

The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat