Oakton pH 5+ Handheld Meter with NIST-Traceable Calibration

Representative image only.
Oakton Item # EW-35613-51
High +/-0.01 pH accuracy at an economical price
  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH and temperature with the press of a button
  • 3 year warranty

PRODUCT OPTIONS

$474.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$39.00 - $405.00USD / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Battery4 AAA (included)
  • Battery life (hr)700
  • Width (cm)8.5725
  • Height (cm)4.28625
  • Width (in)3 3/8
  • Height (in)1 11/16
  • Descriptionmeter only with NIST-traceable Calibration
  • Warranty3 year
3 Year Warranty

Includes

4 AAA batteries.

Key Features

  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH and temperature with the press of a button
  • 3 year warranty

More About this Item

These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.

What's Included: ATC probe, protective rubber boot, and batteries.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat