Oakton PCTSTestr™ 30 pH/Conductivity/TDS/Salinity Waterproof Pocket Tester

Representative image only.
Oakton Item # EW-35634-34
Ultimate tester for pH, conductivity, TDS, salinity and temperature!
  • Combine measurement capabilities of three testers in one convenient pocket-sized unit
  • Up to 3-point pH calibration with NIST or USA auto buffer recognition
  • Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
  • Redesigned leakproof cap functions as a sample, calibration, and hydrating cup
  • Auto-shutoff function conserves battery life
  • IP67-rated housing is waterproof and floats when sensor cap is on
  • 2 year warranty

$269.00USD / Each

$88.50 - $161.00USD / Each

for an additional
$88.50USD / Each
for an additional
$161.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$39.00 - $405.00USD / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Display typeDual-line LCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01 pH
  • pH Accuracy±0.01 pH, ±1 digit
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 pts
  • Min Conductivity0.00 mS/cm
  • Max Conductivity20.00 mS/cm
  • Conductivity Resolution0.1 µS/cm, 1 µS/cm, 0.01 mS/cm
  • Conductivity Accuracy±1% full scale
  • Min TDS0.1 ppt
  • Max TDS10.0 ppt
  • TDS Resolution0.1 ppm, 1 ppm, 0.01 ppt
  • TDS Accuracy±1% full scale
  • Min Salinity0.0 ppt
  • Max Salinity10.0 ppt
  • Salinity Resolution0.1 ppt
  • Salinity Accuracy±1% full scale
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±1.0ºC / ±1.8 ºF
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)200
  • Length (cm)17.8
  • Width (cm)4
  • Height (cm)4
  • Width (in)1 1/2
  • Length (in)7
  • Height (in)1 1/2
  • DescriptionPCTS 30 pH/Conductivity/TDS/Salinity Waterproof Pocket Tester
  • Warranty2 year
Two year warrantyConformite EuropeenneIP 67

Includes

Protective plastic storage case, lanyard, and four AAA batteries.

Key Features

  • Combine measurement capabilities of three testers in one convenient pocket-sized unit
  • Up to 3-point pH calibration with NIST or USA auto buffer recognition
  • Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
  • Redesigned leakproof cap functions as a sample, calibration, and hydrating cup
  • Auto-shutoff function conserves battery life
  • IP67-rated housing is waterproof and floats when sensor cap is on
  • 2 year warranty

More About this Item

Great for water, wastewater, laboratory or plant use, the Oakton PCTS 30 multiparameter tester features long-life pH electrodes with a wide range of sample compatibility. Pin-style conductivity sensors feature stainless steel electrodes for chemical resistance and durability. Replaceable sensor module saves you money by reusing the meter body.
The double junction tester helps you bring the performance of your lab instruments out into the field. Features include large easy-to-read LCD with 2-color backlight, user-adjustable temperature coefficient, self-adjusting TDS and salinity factors, and low-battery indicator. Leakproof cap provides stabilizing base to stand the meter upright.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat