Oakton PC700 Meter w/pH electrode, conductivity/temp probe, and electrode stand

Representative image only.
Oakton Item # EW-35413-00
Easily toggle between pH, mV, conductivity, TDS, and temperature measurements at the touch of a button
  • Space-saving design takes up less room on the bench than other meters
  • Large display lets you see your measurements from across the lab
  • Conductivity and TDS autoranging chooses the best resolution for each measurement
  • 3 year warranty

PRODUCT OPTIONS

$1,204.00USD / Each

$164.50USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Conductivity0 µS
  • Max Conductivity200 mS
  • Conductivity Resolution0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS
  • Conductivity Accuracy±1% FS
  • Min TDS0 ppm
  • Max TDS100.0 ppt at 0.5 factor (200.0 ppt at 1.0 factor)
  • TDS Resolution0.01, 0.1, 1 ppm; 0.01 0.1 ppt
  • TDS Accuracy±1% full-scale
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution0.1°F or °C
  • Temperature Accuracy±0.9°F (±0.5°C)
  • Temperature compensationAutomatic or manual
  • Data Logging (points)100 data sets
  • Power (VAC)110 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Length (in)6 1/8
  • Height (in)2 3/4
  • NIST Calibration IncludedNo
  • Probe IncludedYes
  • DescriptionMeter w/pH electrode, conductivity/temp probe, and electrode stand
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Space-saving design takes up less room on the bench than other meters
  • Large display lets you see your measurements from across the lab
  • Conductivity and TDS autoranging chooses the best resolution for each measurement
  • 3 year warranty

More About this Item

The PC700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance. Dual-display allows you to read both a measured parameter and temperature at a glance. The meter features pH auto-buffer recognition for both USA and NIST buffer sets (1.68, 4.01, 7.00, 10.01 and 12.45 or 1.68, 4.01. 6.18. 9.86, and 12.45), automatic or manual temperature compensation, temperature in °C, display Hold, and water-resistant membrane keypad.

Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants (0.1, 1.0 and 10) are stored in memory for quick and easy retrieval. The convenient pull-out quick reference card keeps keypad functions at your fingertips.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat