Oakton Ion Selective Electrode (ISE), Single-Juntion, Epoxy; Chloride
Representative image only.
Oakton Item # EW-35802-12
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
PRODUCT OPTIONS
$363.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction, solid state
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeChloride
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Single-Juntion, Epoxy; Chloride
  • Warranty1 year
1 Year Warranty
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.