Oakton Grip-Clip Probe Holder for Portable Meters
Representative image only.
Oakton Item # EW-35614-50
Simply clip holder to side of beaker or container for easy measurement
  • Convenient for portable meter measurements with pH and conductovity probes
  • Holds an ATC probe with a 11-/12-mm probe or a 16-mm probe
  • 1 year warranty
$58.65USD / Each
$69.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialABS plastic
  • Compatible WithOakton Portable Meters
  • DescriptionGrip-Clip Probe Holder for Portable Meters
  • Warranty1 year
1 Year Warranty
Key Features
  • Convenient for portable meter measurements with pH and conductovity probes
  • Holds an ATC probe with a 11-/12-mm probe or a 16-mm probe
  • 1 year warranty
More About this Item
Simply clip holder on side of beaker or container to support an ATC probe and a 11-/12-mm or 16-mm probe. Allows for easy, hand-free measurement.