Environmental Express EC300 EC Sensor Head; 2-m Cable
Representative image only.
Environmental Express Item # EW-35660-90
Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
PRODUCT OPTIONS
$291.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeEC Sensor Head
  • Range0 uS/cm to 2000 mS/com
  • MaterialABS plastic and polycarbonate
  • Length (cm)8.5
  • DescriptionEC Sensor Head; 2-m Cable
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
More About this Item
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.