Environmental Express DO300 Optical DO Sensor; 2-m Cable
Representative image only.
Environmental Express Item # EW-35660-96
Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 1 year warranty
PRODUCT OPTIONS
$914.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeOptical DO Sensor with cap
  • Range0 to 20 mg/L
  • Accuracy±2%
  • Materialpolycarbonate, ABS plastic, PMMA, and 316L stainless steel
  • Length (cm)20
  • DescriptionOptical DO Sensor; 2-m Cable
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Includes
DO sensor with cap, calibration bottle, and protective guard.
Key Features
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 1 year warranty
More About this Item
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.