Environmental Express DO200 Field DO Electrode with Integrated Temperature Sensor; 5-m Cable
Representative image only.
Environmental Express Item # EW-35660-62
Enhance the versatility of your Oakton 200 series meters
  • Suitable for use with Oakton 200 series meters and meter kits
  • 6 month warranty
PRODUCT OPTIONS
$722.00USD / Each
Include Meter Today!
SPECS & DESCRIPTION
Specifications & Description
  • ConnectionsBNC and phono
  • Min Dissolved Oxygen0 mg/L
  • Max Dissolved Oxygen20 mg/L
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Length (cm)16.5
  • Diameter (cm)3
  • Length (in)6 1/2
  • DescriptionField DO Electrode with Integrated Temperature Sensor; 5-m Cable
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Includes
DO tip holder and DO tip.
Key Features
  • Suitable for use with Oakton 200 series meters and meter kits
  • 6 month warranty
More About this Item
For your convenience, cables, electrodes, and replacement parts are available for your Oakton meters.