Oakton Conductivity Probe Storage Solution; 500 mL
Representative image only.
Oakton Item # EW-00652-80
Keep probes in working order
  • Ideal for storing and conditioning all glass and epoxy conductivity probes
$41.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)500
  • Volume (oz)16
  • DescriptionConductivity Probe Storage Solution
1 Year Warranty
Key Features
  • Ideal for storing and conditioning all glass and epoxy conductivity probes
More About this Item
Conductivity Probe Storage Solution is ideal for storing and conditioning your conductivity cells. It preserves the probe's cleanliness, allows immediate use, and reduces contamination that can occur with storing probe in water only.
Please Note: This item does not have a certificate of analysis. The Lot# listed is for the manufacturers quality control tracking.