Oakton Buffer Pack; 60 mL of each pH 1.68, 4.01, 7.00, 10.01, and 12.46

Representative image only.
Oakton Item # EW-00653-14
Save over 25% on pack compared to ordering separately
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Perfecty sized for portable needs
  • 1 year warranty

PRODUCT OPTIONS

$155.00USD / Pkg of 5
Storage & Cleaning Solutions…

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Kit Values1.68, 4.01, 7.00, 10.01, and 12.46
  • Volume (mL)60
  • DescriptionBuffer Pack; 60 mL of each pH 1.68, 4.01, 7.00, 10.01, and 12.46
  • Warranty1 year
1 Year Warranty

Includes

one, 60-mL plastic bottle of each pH solution: 1.68, 4.01, 7.00, 10.01, and 12.46.

Key Features

  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Perfecty sized for portable needs
  • 1 year warranty

More About this Item

• Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
• Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
• Store buffers away from direct sunlight.

• Color coded for quick visual reference:
- pH 1.68 buffer in clear
- pH 4.01 buffer in red
- pH 7.00 buffer in yellow
- pH 10.01 buffer in blue
- pH 12.46 buffer in clear

Convenient 60-mL bottles come in a pack of five—ideal for field use. Grab one when you're on the go!

Contains all "USA" pH calibration buffer values in one, convenient, pack.

To obtain a Certificate of Analysis on each item within the pack, please visit each individual items' page and enter the lot number in the space provided. For example, type part number 00654-00 into the search box at the top web page, then scroll down on the page to the CERTIFICATE OF ANALYSIS box, and enter the lot number in the space provided.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat