Oakton Buffer Pack; 10 x 500 mL of each pH 4.01, 7.00, and 10.01

Representative image only.
Oakton Item # EW-05942-11
Save over 25% on pack compared to ordering separately
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Bundle and save even more!
  • 2 year warranty

PRODUCT OPTIONS

$405.00USD / Pkg of 30
New Double Junction Waterproof Oakton pH Testr 50

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Kit Values4.01, 7.00, and 10.01
  • Volume (mL)500
  • DescriptionBuffer Pack; 10 x 500 mL of each pH 4.01, 7.00, and 10.01
Two year warranty

Includes

Ten, 500-mL plastic bottles of each pH solution: 4.01, 7.00, and 10.01—Thirty bottles total!

Key Features

  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Bundle and save even more!
  • 2 year warranty

More About this Item

• Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
• Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
• Store buffers away from direct sunlight.

• Color coded for quick visual reference:
- pH 4.01 buffer in red
- pH 7.00 buffer in yellow
- pH 10.01 buffer in blue

The most common "USA" pH calibration buffer values—in one convenient pack!

To obtain a Certificate of Analysis on each item within the pack, please visit each individual items' page and enter the lot number in the space provided. For example, type part number 00654-00 into the search box at the top web page, then scroll down on the page to the CERTIFICATE OF ANALYSIS box, and enter the lot number in the space provided.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat