Oakton Accumet pH Test Paper, 0 to 13 Indicator Roll
Representative image only.
Oakton Item # EW-35850-08
Double-wide continuous paper roll is glove friendly
  • 3 month warranty
$25.60USD / Pkg
Include Accessories Today!
Oakton pH Indicator Strips
$30.50USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points0 to 13 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit150
  • DescriptionAccumet pH Test Paper, 0 to 13 Indicator Roll
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Oakton pH indicator paper features a 1.0 pH unit interval across the entire range. Packed in a plastic dispenser with an extra-large color comparator chart, the test paper features vivid and easily distinguished colors provide an instant result. Paper roll measures 1/2" wide x 50-ft long (1.3 cm x 15.2 m), enough for approximately 150 tests.