This item is now discontinued. We recommend the following as suggested replacements: EW-81660-16
NIOSH N99 Particulate Respirator with valve, Metal Nose Seal; 12/box
Representative image only.
Mfr # 1914 – Item # EW-78901-27
NIOSH approved and CDC listed particulate respirator
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NOISH 42CFR84
  • Exhalation valve keeps you cool
SPECS & DESCRIPTION
Specifications & Description
  • RatingN-99
  • NIOSH ApprovalNOISH 42CFR84
  • ComplianceNIOSH approved, CDC listed
  • DescriptionNIOSH N99 Particulate Respirator with valve, Metal Nose Seal
Key Features
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NOISH 42CFR84
  • Exhalation valve keeps you cool
More About this Item
NIOSH approved particulate respirators protect against solid and liquid aerosols that do not contain oil. It features a flexible metal nose clip seal with stapled straps. Optional exhalation valve keeps you cool.