This item is now discontinued. We recommend the following as suggested replacements: EW-40111-04
NIOSH N95 Particulate Respirator with valve, Nose Bridge Seal; 12/box
Representative image only.
Mfr # 15144 – Item # EW-78901-25
NIOSH approved and CDC listed particulate respirator
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NIOSH 42CFR84
  • Exhalation valve keeps you cool
SPECS & DESCRIPTION
Specifications & Description
  • RatingN-95
  • NIOSH ApprovalNOISH 42CFR84
  • ComplianceNIOSH approved, CDC listed
  • DescriptionNIOSH N95 Particulate Respirator with valve, Nose Bridge Seal
Key Features
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NIOSH 42CFR84
  • Exhalation valve keeps you cool
More About this Item
NIOSH approved particulate respirators protect against solid and liquid aerosols that do not contain oil. The ergonomic shape with welded straps gives you the perfect nose bridge seal. Optional exhalation valve keeps you cool.