Cole-Parmer Essentials Nickel/SS Lab Spatulas with 1.13" Spoon x 1.63" Flat Blunt Ends; 3/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06369-16
Get precise sampling of powders or salts
  • Easy-to-clean finish
  • Choose the right end style for your needs
PRODUCT OPTIONS
$40.00USD / Pkg of 3
BUY MORE & SAVE
  • 1-5$40.00USD / Pkg of 3
  • 6-29$36.00USD / Pkg of 3
  • 30-119$34.00USD / Pkg of 3
  • 120+$32.00USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)8 7/8
  • Overall Length (cm)22.54
  • Material304 Stainless Steel, Nickel
  • SterileNo
  • DescriptionNickel/SS Lab Spatulas with 1.13\" Spoon x 1.63\" Flat Blunt Ends; 3/Pk
Key Features
  • Easy-to-clean finish
  • Choose the right end style for your needs
More About this Item
Nickel/stainless steel spatulas come in a variety of end styles. All spatulas have a glazed finish, except for models 06369-05, and -16 which have a mirror finish.