Modular stainless steel drying rack, 32 assorted color pegs

Representative image only.
Item # EW-67035-02
  • Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds!
  • Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable.
  • Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.
  • Racks come with 6"L pegs, SS drain tube, and wall mounting bracket. Order add-on accessories, plugs, and additional pegs separately.

PRODUCT OPTIONS

$681.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)4
  • Width (in)24
  • Height (in)30
  • Length (mm)10.2
  • Width (mm)610
  • DescriptionModular stainless steel drying rack, 32 assorted color pegs

Key Features

  • Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds!
  • Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable.
  • Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.
  • Racks come with 6"L pegs, SS drain tube, and wall mounting bracket. Order add-on accessories, plugs, and additional pegs separately.

More About this Item

Assorted peg colors include yellow, orange, pink, red, green, light blue, dark blue, lilac, and purple.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat