Mitutoyo 950-315 Electronic/Digital Protractor
Representative image only.
Mitutoyo Mfr # 950-317 – Item # EW-97230-00
  • 1 year warranty
$390.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min range (degrees)0
  • Max range (degrees)360
  • Resolution0.1°
  • Accuracy±0.1° at level and plumb.
  • DescriptionElectronic/Digital Protractor
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Digital protractor measures angles in the full 360°> range. Resolution is ±0.1°>; accuracy is ±0.1°> at level and plumb. Zero setting and hold. Operates on one 9 V battery (included).