Mettler Toledo Hydrofluoric pH Electrode; S7
Representative image only.
Mettler Toledo Mfr # 51343176 – Item # EW-35902-10
Expertly-engineered probes to cover a wide range of applications
  • Cable not included⌡Special HF resistant glass membrane
PRODUCT OPTIONS
$635.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSolutions containing concentrations up to 1 g/L HF acid
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHF - Hydrofluoric acid applciations
  • Min pH (pH)1
  • Max pH (pH)11
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionpH Electrode; S7
Key Features
  • Cable not included⌡Special HF resistant glass membrane
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available.⌡Appropriate for samples that contain hydrofluoric acid (up to 1 g/L).