Mettler Toledo Flex-Micro pH Electrode; BNC with 3.3 ft Cable

Representative image only.
Mettler Toledo Mfr # 51343164 – Item # EW-35901-46
Expertly-engineered probes to cover a wide range of applications
  • ARGENTHAL™ reference system

PRODUCT OPTIONS

$700.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ApplicationsSmall volumes and containers
  • Electrode Body MaterialEpoxy
  • Reference JunctionPorous PTFE
  • Reference ElectrolyteNon-refillable gel
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • GuardYes
  • ATC Element
  • ConnectorBNC
  • Diameter (mm)6
  • Length (mm)180
  • Cable Length (m)1
  • DescriptionpH Electrode; BNC with 3.3 ft Cable

Key Features

  • ARGENTHAL™ reference system

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat