Mettler Toledo 18552 Capillary Tubes, 1.3 to 1.5-mm dia.; 150/PK
Representative image only.
Mettler Toledo Mfr # 18552 – Item # EW-03014-62
Precision accuracy of ±0.5°C provides high reproducibility of results
  • Meet GLP requirements with optional printer
  • Digital display eliminates interpretation errors
  • 3 month warranty
$159.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCapillary tubes, 1.3 to 1.5-mm dia., 150/pk.
  • Warranty3 month
3 Month Warranty
Key Features
  • Meet GLP requirements with optional printer
  • Digital display eliminates interpretation errors
  • 3 month warranty
More About this Item
This automatic melting point apparatus is ideal for determining sample purities for quality control, educational, and research and development applications. Unit measures the reflection of incident light in the crystalline state and the increase in light transmittance during the melting process. Operation is simple — insert the test tube containing one to three mg of sample into the instrument, select the starting temperature and heating rate (user selectable between 0.1°C and 10°C per minute in steps of 0.1°C), and the visual and audible signals will indicate when the melting point is acheived.
The operator interface provides graphic display aids making set up and programming simple. The large LCD displays temperature readings to 0.1°C.