Meiji Techno MX4300L Economical Compound Microscope Trinocular; 4x/10x/40x/1000x; 110-240 V
Representative image only.
Meiji Techno Mfr # MT4300L – Item # EW-48400-03
Infinity-corrected optical system design for high-image quality, contrast, and flatness of field
  • Smooth-operating, ball-bearing quintuple nosepiece provides effortless objective changes
  • Wide ceramic-coated stage for smooth exchange of specimens
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification4x/10x/40x/100x
  • Maximum Magnification1000x
  • Head ConfigurationTrinocular
  • ObjectivesSemi-plan
  • Eyepiece Diameter (mm)23.2
  • Eyepiece Magnification10x
  • Stage Dimensions171 x 115 mm
  • Illuminator(s)LED
  • Power (VAC)100 to 240
  • DescriptionEconomical Compound Microscope Trinocular; 4x/10x/40x/1000x; 110-240 V
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Smooth-operating, ball-bearing quintuple nosepiece provides effortless objective changes
  • Wide ceramic-coated stage for smooth exchange of specimens
  • Limited Lifetime Warranty
More About this Item
Economical Compound Microscopes
These economical microscopes with S. Plan optics are ideal for education and research laboratory applications. Feature LED illumination and comfortable siedentopf viewing heads with 23.2-mm eyetubes, and F.N. 20, KHW10X widefield eyepieces.
What’s included: Abbe 1.25 N.A. condenser in dovetail mount, 4x/10x/40x/100x oil objectives, S. Plan objectives, auto-voltage sensing power supply, 6-ft detachable power cord, green filter, dust cover, and immersion oil.