DisplayWVGA color graphics LCD (800 x 480 pixels), user-selectable color schemes; viewable in external environments
Battery Life12 hours (typical)
Battery1 rechargeable lithium-ion (included)
Length (in)11 3/8
Width (in)7 1/2
Height (in)2 1/4
Power110 VAC for recharging batteries
DescriptionDual-Channel Advanced Step Time Domain Reflectometer for Power Applications
Warranty2 years
Includes
Retractable sheath fused test leads (2 pairs), TraceXpert software with USB cable, carrying case, Li-ion rechargeable battery, and AC/CD power supply and charger.
Key Features
Ensure the most effective parameters are selected with AUTO ranging option
Dual-channel, dual-trace, and dual-cursor capabilities provide instant distance between points
Use trace comparison to closely examine conditions between different traces
Store up to 100 trace measurements
Included TraceXpert software assists with downloading, reporting, and uploading
Can be used on energized circuits up to 600 V CAT IV
Step TDR technology detects near end faults
Enhanced features—distance dependent gain, autofind, and findend—provide easier fault detection
Assign a name to all saved traces
Six additional set color schemes available to help improve display viewing
2 year warranty
More About this Item
These advanced dual-channel TDRs help locate cable faults for responsive or routine maintenance; quality control checks in manufacturing, shipping, and inventory management; and performance checks on cables in hard-to-reach or hazardous locations. The Auto option aids in rapid diagnosis of the TDR trace; manual override allows fine tuning and identification of difficult to determine faults.
Easily set up and view traces on a large, high-resolution color WVGA display. Directional control buttons and soft keys make it easy to use. Extra high resolution, backlight, and user definable tones and color provide vibrance to the graphical display, making it easier to identify events on the trace. Use trace comparison feature to closely examine conditions between different traces.
The TDRs store up to 100 traces in the internal memory. Recall the stored traces to the display for analysis or compare with an active display to help in fault location. Use the included TraceXpert software to download to a PC via the USB port for reporting and analysis. An IP54-rated case protects meter from dust and rain.
This step TDR features a built-in 600 V CAT IV input protection filter for connecting to known live lines. Step TDR technology injects the signal at full strength until a disturbance is detected—perfect for detecting near end faults standard pulse TDRs miss. Additionally, it features Distance Dependent Gain (DDG) for a more even representation of the relative attenuation at all points along the trace. Automatically positions the cursor to the first major event along the result trace using the AutoFind feature; automatically detect the end of the cable with the FindEnd feature. Use the trace tagging feature to assign a name (up to 32 alphanumeric characters) to each saved trace. Stored traces may be recalled and compared to an active display to aid in fault location.