Masterflex Ultrasonic Flow Sensor Communications Package
Representative image only.
Masterflex Item # EW-33501-96
Make the most of your Masterflex flowmeters with these replacement parts and other accessories
  • Optimize performance of your flowmeter with these accessories
  • Provides an easy connection to the flow sensor to facilitate calibration
  • Software makes it easy to finely tune the flow sensor for the flow range that the application needs
  • Enables data to be recorded / captured for real-time observation using a connected computer or download to review at a later time
PRODUCT OPTIONS
$1,256.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With33501-13, 33501-14, 33501-15, 33501-16, 33501-17, 33501-18, 33501-24, 33501-25, 33501-26, 33501-35, 33501-36, 33501-73
  • ConnectionsOne USB connects to computor for power and data, 2nd USB for addtional power, M12 attaches to Masterflex Flow Sensor
  • DescriptionMasterflex Ultrasonic flow sensor communications package
Includes
Specially adapted cable (has one USB for both data and power and a second USB for additional power - if needed - on one side and on the other is a M12 connector for attaching to the Masterflex Flow Sensor) ; and software on a USB drive
Key Features
  • Optimize performance of your flowmeter with these accessories
  • Provides an easy connection to the flow sensor to facilitate calibration
  • Software makes it easy to finely tune the flow sensor for the flow range that the application needs
  • Enables data to be recorded / captured for real-time observation using a connected computer or download to review at a later time
More About this Item
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
The USB data converter is used to communicate via the serial interface to the Masterflex Ultrasonic Flow Sensor using a computer. The same data cable can used on any of the Masterflex Flow Sensors.
The software / data cable enable simple / fine adjustment of the flow sensors to best match it to a specific task.
To assist in problem solving, one can view real-time data in a graphical display when connected to a computer or save data points for later analysis.
Allows one to use the sensor on a different tubing formulation or wall thickness if desired.
Supported operating systems:
- Win98, Win2000, Win XP, Win7, Win8, Win10
Screen resolution:
- 1024 x 640 or higher
Interface:
-1 USB port
Processor:
- Pentium 700 MHz or higher
RAM:
- 256 MB or higher
Software (needed by user - not provided):
- Excel (for the management of the measurement data)