Masterflex Two-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
Representative image only.
Masterflex Item # EW-30503-04
Easily connect to rigid tubing
$256.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeCompression
  • DescriptionTwo-Way Stopcock for Glass and Rigid Tubing, 10 mm tube OD
More About this Item
Stopcocks have wetted parts constructed of PFA. An O-ring made of Viton® protects the glass tube when you tighten the compression nut. Stopcocks also accept rigid tubing such as PTFE.
Available in two-way and three-way styles.