Masterflex Spray Gun, PTFE for DI Water, 1/2" NPT(F); 1/Ea
Representative image only.
Masterflex Item # EW-98515-03
No splashing
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
PRODUCT OPTIONS
$186.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeNPT(F)
  • Wetted MaterialsPTFE
  • DescriptionSpray Gun, PTFE for DI Water; 1/2"
Key Features
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
More About this Item
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.