Masterflex Solenoid Pinch Valve, Normally-Closed, for L/S 17 tubing; 12 VDC
Representative image only.
Masterflex Item # EW-98306-24
Specifically designed for Masterflex® pump tubing
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
PRODUCT OPTIONS
$175.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypePinch Valve for Tubing
  • Power (VDC)12
  • Watts10
  • Response30 to 50 msec
  • Masterflex Tube SizeL/S 17
  • Flow PatternNormally Closed
  • DescriptionSolenoid Pinch Valve; for use with L/S 17 tubing, normally closed, 12 VDC
Key Features
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
More About this Item
Easily control the flow through your Masterflex tubing.