Masterflex Single-Use Tubing Pinch Clamp, Nylon; 1/Ea
Representative image only.
Masterflex Item # EW-19999-99
Swing-away lockdown nut provides ease of use
  • Higher tensile strength than polysulfone—less likely to break
  • Offers quick release from locked position
  • Ideal for biopharma applications that require gamma irradiation or autoclave cycles
$34.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Minimum ID (in)0.187
  • Maximum ID (in)0.5
  • MaterialNylon
  • DescriptionNylon Tubing Pinch Clamp for Tubing and Hose
Key Features
  • Higher tensile strength than polysulfone—less likely to break
  • Offers quick release from locked position
  • Ideal for biopharma applications that require gamma irradiation or autoclave cycles
More About this Item
This clamp accepts tubing from 0.187 to 0.500" inner diameters. The recommended wall thickness is 0.085 to 0.200". Tubing materials that can be used include silicone, TPE TPV, PVC, and other rubbers. The maximum hardness for correct functioning of the clamp is 70 Shore A. May be installed on existing tube assemblies without disrupting the process fluid path.