Masterflex Flowmeter Tripod Base for One, Two, or Three Meters
Representative image only.
Masterflex Item # EW-03226-30
Easily mount flowmeters on tripod for easy viewing
  • Constructed of durable acrylic
PRODUCT OPTIONS
$67.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlowmeter Tripod Base for One, Two, or Three Meters
Key Features
  • Constructed of durable acrylic
More About this Item
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.