Masterflex Flow-Through Tubing Weight; Stainless Steel
Representative image only.
Masterflex Item # EW-07570-04
Keep your samples flowing and tubing from curling
  • Choose between PTFE or stainless steel construction
PRODUCT OPTIONS
$112.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Minimum ID (in)0.1875
  • Maximum ID (in)0.25
  • Minimum ID (mm)6.8
  • MaterialStainless Steel
  • DescriptionStainless steel flow-through tubing weight.
Key Features
  • Choose between PTFE or stainless steel construction
More About this Item
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.