Masterflex Dispensing Gun PTFE 1/2"npt (Spray Gun Valves (actuated))
Representative image only.
Masterflex Item # EW-98515-07
No splashing
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
PRODUCT OPTIONS
$208.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeNPT(F)
  • Wetted MaterialsPTFE
  • DescriptionDispensing Gun PTFE 1/2"npt (Spray Gun Valves (actuated))
Key Features
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
More About this Item
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.