Mark-10 ES10 Test Stand, Lever-Operated, 100 lbF / 500 N
Representative image only.
Mark-10 Mfr # ES10 – Item # EW-59846-16
Ideal for low force applications
  • Small footprint well suited for limited brench space
  • Rugged design offers years of service
  • 3 year warranty
PRODUCT OPTIONS
$660.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)100
  • Weight (lbs)10
  • DescriptionTest stand, Lever Operated, 100 lb
  • Warranty3 year
3 Year Warranty
Key Features
  • Small footprint well suited for limited brench space
  • Rugged design offers years of service
  • 3 year warranty
More About this Item
Test stands produce more repeatable results than handheld testing. Several different sizes, loading methods, and capacities are available. All stands are ruggedly designed for many years of service in laboratory or production environments.