M-System MDP-24-1 Lightning & Surge Protector For 4-20mA And Pulse Signal
Representative image only.
M-System Mfr # MDP-24-1/UL – Item # EW-68467-67
Don’t let lightning take systems down
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed
  • 1 year warranty
PRODUCT OPTIONS
$96.26USD / Each
$113.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLightning & Surge Protector For 4-20mA And Pulse Signal
  • Warranty1 Year
1 Year Warranty
Key Features
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed