LaMotte DC-1500 Single Test Colorimeter Kit , Phosphate

Representative image only.
LaMotte Mfr # RBY 3242 – Item # EW-99562-22
Single test, direct-reading colorimeters feature microprocessor technology
  • Analyzer features automatic wavelength selection for quick testing
  • 1 year warranty

PRODUCT OPTIONS

$862.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeLCD
  • Light SourceLED
  • Detector TypeSilicon Photodiode
  • Test ParametersPhosphate
  • Number of Tests Included100
  • Data LoggingYes
  • Battery1 9V (included)
  • Length (in)6 1/2
  • Width (in)3 1/4
  • Height (in)2 1/2
  • DescriptionSingle Test Colorimeter Kit , Phosphate
  • Warranty1 year
1 Year Warranty

Includes

analyzer, glass sample vials, plastic water bottle, tablet crusher, test-specific reagent, foam-lined carrying case, and one 9 V battery.

Key Features

  • Analyzer features automatic wavelength selection for quick testing
  • 1 year warranty

More About this Item

Auto-zeroing and a hinged light cover make these meters easy to use. Ergonomic design fits in your hand comfortably. Water-resistant design protects your meter from damage. Download data to a computer or printer via the optional RS-232 interface.
What's included: analyzer, glass sample vials, plastic water bottle, tablet crusher, test-specific reagent, foam-lined carrying case, and one 9 V battery.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat