Labnet D1301 AccuBlock™ Dry Block Heater, Single; 120 VAC

Representative image only.
Labnet Mfr # D1301 – Item # EW-51401-18
Precise temperature regulation up to 150°C ensures accurate results
  • Single, dual, and four block configurations available
  • Dual digital displays
  • External probe available for better traceability

PRODUCT OPTIONS

$377.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Length (")12 7/16
  • Number Of Blocks1
  • Uniformity (° C)0.2
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Power (VAC)120
  • DescriptionLabnet D1301 AccuBlock™ Dry Block Heater, Single; 120 VAC

Key Features

  • Single, dual, and four block configurations available
  • Dual digital displays
  • External probe available for better traceability

More About this Item

AccuBlock Digital Dry baths are available in single, dual, and four block configurations. Ideal in a variety of applications including determining melting and boiling points, cell cultures, enzyme reactions and many more. Built of anodized, high grade non-porous aluminum, these units extremely durable and come with USB connectivity included.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat