Labconco FlaskScrubber® Freestanding Vantage® Glassware Washer with Window, 230 VAC, 1-Phase

Representative image only.
Labconco Mfr # 422101010 – Item # EW-88964-24
CleanPoint™ Technology measures conductivity and bypasses unnecessary pure water rinses
  • SpeedVent™ Technology shortens drying time by venting residual heat from the washer
  • HEPA-filtered drying provides particulate-free forced air to quickly remove moisture
  • Robust heating
  • Water temperature reaches 93° C to remove stubborn contaminants
  • Lower spindle rack included with 36 spindles

PRODUCT OPTIONS

$19,890.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Water Consumption (Gal)3.4
  • Power (VAC)230
  • Power (Hz)50/60 Hz, 1-phase
  • Washing Temperature (° F)199.4
  • Washing Temperature (° C)93
  • Shipping Weight (lbs)294
  • DescriptionFreestanding Vantage® Glassware Washer with Window, 230 VAC, 1-Phase
Two year warranty

Key Features

  • SpeedVent™ Technology shortens drying time by venting residual heat from the washer
  • HEPA-filtered drying provides particulate-free forced air to quickly remove moisture
  • Robust heating
  • Water temperature reaches 93° C to remove stubborn contaminants
  • Lower spindle rack included with 36 spindles

More About this Item

FlaskScrubber® Vantage® glassware washers contain powerful dual pumps to recirculate water and a separate drain pump to reduce the potential for cross contamination. The peristaltic pumps act as automatic detergent and rinse aid dispensers by automatically drawing a programmed amount of liquid detergent and rinse aid during the wash and rinse cycles. The steam generator produces hot vapor to penetrate and soften dried contaminants for more effective cleaning. The CleanWorks™ OS operating system makes control simple. The progress bar displays segment progression of a running program from the home screen. Up to 200 programs can be created and stored. The data can be viewed as a table or graph. Table view can be customized to show up to 6 data columns. Results can be exported via USB to backup programs and files.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat