Labconco FlaskScrubber 4595400 Base Stand for Freestanding FlaskScrubber or SteamScrubber
Representative image only.
Labconco Mfr # 4595400 – Item # EW-98963-50
Optimize performance of your Labconco glassware washer
  • Adds more cleaning options to your washer
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBase stand for Freestanding FlaskScrubber or SteamScrubber
  • Warranty2 year
Two year warranty
Key Features
  • Adds more cleaning options to your washer
  • 2 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Base stand is made of 304 stainless steel and raises the washer 17" for easier loading and unloading. Unit incorporates one storage cabinet with hinged door for storage. Comes fully assembled with four leveling feet.