Labconco 4591900 Drain Water Cooling Kit; 115V, 60Hz
Representative image only.
Labconco Mfr # 4591900 – Item # EW-98963-55
Optimize performance of your Labconco glassware washer
  • Adds more cleaning options to your washer
  • 1 year warranty
PRODUCT OPTIONS
$975.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionDrain water cooling kit, 115VAC 60Hz
  • Warranty1 year
1 Year Warranty
Key Features
  • Adds more cleaning options to your washer
  • 1 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Cooling kit mixes hot discharged water with cold tap water to reduce the temperature before releasing into the drain pipes. Unit is perfect for locations where plumbing cannot tolerate higher temperature water. Comes complete with plumbing components, solenoid valve, and electrical connections.