LabChem Water, Deionized, ASTM Type II, 4 L

Representative image only.
LabChem Mfr # LC267504 – Item # EW-80055-06
Quality and delivery you can count on every time!
  • Water meets ASTM Type II standards
  • Water comes in 4 L size and packaged in a poly bottle

PRODUCT OPTIONS

$40.30USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeASTM Type II
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)4000
  • Formula Weight18.02
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)60
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, Deionized, ASTM Type II; 4 L

Key Features

  • Water meets ASTM Type II standards
  • Water comes in 4 L size and packaged in a poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat