KN95 Respirator Mask with Non-Woven Fabric, >99% BFE (2.5 µm), >95% PFE (0.5 µm) ; Box of 10
Representative image only.
Mfr # MASK2-K95 – Item # EW-78901-22
Five layers of fabric provide good particle filtration
  • Curved shape for easy breathing
  • Comfortable to wear—soft ear loops and adjustable nose bridge strip
  • Easy to put on and take off
  • Manufactured in an FDA-registered facility
$33.50USD / Box of 10
BUY MORE & SAVE
  • 1-4$33.50USD / Box of 10
  • 5+$30.15USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • ColorWhite
  • DescriptionKN95 Respirator Mask with Non-Woven Fabric, >99% BFE (2.5 µm), >95% PFE (0.5 µm) ; Box of 10
Key Features
  • Curved shape for easy breathing
  • Comfortable to wear—soft ear loops and adjustable nose bridge strip
  • Easy to put on and take off
  • Manufactured in an FDA-registered facility
More About this Item
This disposable respirator mask is lightweight and comfortable with soft ear loops, an adjustable nose bridge, and curved design. The five layers provide a particle filtration efficiency of ≥95% and bacterial filtration efficiency of ≥99%. There are two non-woven fabric layers, two melt blown fabric layers, and one hot air cotton layer. The mask is non-sterile and latex-free.