Cole-Parmer TELOS® SPE Microplate, 10 µm PE frit, fixed wells; 1/pk
Representative image only.
Cole-Parmer Item # EW-06478-31
Pack wells with custom sorbents
  • Optimized for the sample processing and extraction of small volume fluids
  • Use with vacuum or positive pressure
PRODUCT OPTIONS
$64.50USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Fritted Disk10 µm
Key Features
  • Optimized for the sample processing and extraction of small volume fluids
  • Use with vacuum or positive pressure
More About this Item
Individual well is designed to allow maximum interaction between the analyte and sorbent and ensure elution volumes are kept to a minimum. Fill with your own custom sorbent to meet your application. Each well is fitted with a polyethylene frit.