Cole-Parmer TELOS® neo™ MicroPlate™ Method Development Kit, 10mg
Representative image only.
Cole-Parmer Item # EW-06474-19
Optimize performance of your Cole-Parmer® TELOS® neo™ MicroPlate™
  • Choose from a range of accessories to enhance performance of your MicroPlate
  • Includes 20 each of TELOS neo PRP, PCX, WCX, PAX, and WAX 10 mg wells
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithKinesis TELOS® neo™ MicroPlate™ loose wells
  • DescriptionMethod Development Kit, 10mg
Key Features
  • Choose from a range of accessories to enhance performance of your MicroPlate
  • Includes 20 each of TELOS neo PRP, PCX, WCX, PAX, and WAX 10 mg wells
More About this Item
Required components, tools, and seals for your SPE microplates.
Build your own method development plate to suit your application. Method development kit contains 20 x 10 mg wells of TELOS neo PRP, PCX, WCX, PAX, and WAX; base plate; base plate sealing strips; and well removing tool.