Cole-Parmer TELOS® Ion Exchange SPE Microplate, NR4+ (SAX), 100 mg sorbent, fixed wells; 1/pk
Representative image only.
Cole-Parmer Item # EW-06477-78
Optimized for sample processing and extraction of small volume biological fluids
  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
  • Applications: Ion exchange
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)100
  • Column Particle Size (µm)50
  • End CapNo
  • ApplicationsIon exchange
  • SorbentSAX
Key Features
  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
  • Applications: Ion exchange
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE fixed 96-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. C4 (WP) sorbents with average pore size of 300 Å are ideal for compounds with molecular weight >10,000.