Cole-Parmer VapLock™ Basic Fittings Kit for 10-Port Waste Manifold, Polypropylene
Representative image only.
Cole-Parmer Item # EW-12017-22
Optimize performance of your Spex® VapLock™ VaporSafe™ systems
  • Extend the life of your solvent collection systems with these replacement parts and accessories
PRODUCT OPTIONS
$73.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • ColorMixed
  • Compatible WithKinesis KX VaporSafe™ Solvent Waste Systems and Bottle Caps
  • DescriptionBasic Fittings Kit for 10-Port Waste Manifold
Includes
PP filter adapter: 1/4" NPT(M) x 1/2" NPT(M) PP plugs: (3) 1/4" NPT(M) and (1) 1/2" NPT(M) PTFE plugs: (6) 1/4"-28 UNF(M) PP adapters for semi-rigid tubing: (2) 2.2 mm OD, (1) 2.5 mm OD, (2) 1/16" OD, and (3) 1/8" OD PP barbed adapters with 1/4" NPT(M) connection for soft tubing: (1) 1/8" ID, (1) 3/16" ID, (3) 1/4" ID, and (1) 3/8" ID PP barbed Y connector: 1/4" ID 5 feet of 1/8" OD PTFE extension tubing with union connections.
Key Features
  • Extend the life of your solvent collection systems with these replacement parts and accessories
More About this Item
Replacement parts, fittings, and other items for your KX VaporSafe solvent collection systems.