Cole-Parmer KX+ LCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk
Representative image only.
Cole-Parmer Item # EW-98701-76
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • 0.3 mL vial constructed of durable polypropylene
  • PTFE/silicone septum
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)0.3
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Cap TypeShort Thread Screw
  • Cap Diameter (mm)9
  • Septum TypePTFE/silicone
  • SterileNo
  • Special FeaturesLimited-Volume
  • DescriptionLCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • 0.3 mL vial constructed of durable polypropylene
  • PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Limited-volume vials are excellent for handling small-volume samples while delivering low residual volume (<8 µL for 300 µL vials; ,80 µL for 700 µL vials). Polypropylene (PP) is ideal for samples where glass is not appropriate; as with samples that adhere to glass.