Kinesis 384-well Collection Plate, Glass Lined PP, Square U-Bottom, 120µL; 10/pk
Representative image only.
Kinesis Item # EW-98700-08
Glass lining provides excellent chemical resistance
  • Constructed of glass-lined polypropylene
  • 384-well, U-bottom, 120 µL
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells384
  • Well Volume (µL)120
  • Well ShapeSquare, U bottom
  • MaterialGlass-lined polypropylene
  • Description384-well Collection Plate, Glass Lined PP, Square U-Bottom, 120µL; 10/pk
Key Features
  • Constructed of glass-lined polypropylene
  • 384-well, U-bottom, 120 µL
More About this Item
Collection plates are available with three well shapes: V-bottom wells let you completely retrieve well content; U-bottom wells optimize coating and washing; and flat wells maximize light transmission.