Keck Metal Standard Taper Clip Size 24 6/Pk
Representative image only.
Keck Mfr # MAKE-UP – Item # EW-06836-15
Nickel-plated steel clips have superior chemical and temperature resistance
  • 1 year warranty
PRODUCT OPTIONS
$90.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialNickel-plated Steel
  • Clamp TypeGlassware, Standard Taper, Joint Size 24
  • DescriptionMetal Standard Taper Clip, Size 24, 6/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These metal taper clips securely hold conical joints under pressure. Nickel-plated steel taper clips feature superior chemical and temperature resistance—withstand temperatures to 932°>F (500°>C).