Jenway Genova Plus Life Science Spectrophotometer, DNA/RNA/Oligo/Protein; 115-230 VAC

Representative image only.
Jenway Item # EW-83070-00
Dedicated to life science applications—preprogrammed for DNA, RNA and protein analysis
  • DNA purity scan applies across the entire wavelength range
  • Icon-driven software and easy-to-use navigation system provide intuitive usability
  • Autologging capabilities enable repeat measurements of the same sample with a selected time interval
  • Minimal footprint with large graphical display
  • Save methods and results to USB memory stick
  • 3 year warranty

$6,538.00USD / Each

$1,405.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Don't forget to order accessories for your spectrophotometer

SPECS & DESCRIPTION

Specifications & Description

  • Source LampXenon
  • Bandwidth5.0 nm
  • Min Wavelength (nm)198
  • Max Wavelength (nm)1000
  • Wavelength Accuracy±2.0 nm
  • Wavelength Reproducibility±0.5 nm
  • Min Photometric - Transmittance (%T)0
  • Max Photometric - Transmittance (%T)199.9
  • Min Photometric - Absorbance (A)-0.3
  • Max Photometric - Absorbance (A)2.5
  • Photometric Accuracy±0.01 A At 1.0 A; ±1%T (0-100%T)
  • Concentration0-9999
  • OutputUSB, RS-232, and analog
  • Display TypeLCD
  • Width (cm)27.5
  • Height (cm)22
  • Depth (cm)40
  • Width (in)10 51/64
  • Height (in)8 45/64
  • Depth (in)15 45/64
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • DescriptionLife Science Spectrophotometer; DNA/RNA/Oligo/Protein; 115 230 VAC
  • Warranty3 year
3 Year WarrantyUL Listed to Canadian safety standards.Conformite Europeenne

Includes

Micro-cuvette holder, 10 x 10 mm cuvette holder, 100 disposable micro-cuvettes, USB memory stick, and power supply.

Key Features

  • DNA purity scan applies across the entire wavelength range
  • Icon-driven software and easy-to-use navigation system provide intuitive usability
  • Autologging capabilities enable repeat measurements of the same sample with a selected time interval
  • Minimal footprint with large graphical display
  • Save methods and results to USB memory stick
  • 3 year warranty

More About this Item

The versatile Genova Plus spectrophotometer has all the features of a standard spectrophotometer plus features that are dedicated to life science analysis. Measure DNA concentrations and purity ratios using wavelengths recorded at 260, 280 and 230 nm, with an optional correction at 320 nm. Preprogrammed methods for measuring Bradford, Lowry, Biuret, and Bicinchoninic Acid (BCA) assays minimize errors.

The OD measurement mode measures optical density at 600 nm for cell harvesting. The purity scan across the entire wavelength range from 198 to 1000 nm displays any distorted peaks enabling impurities to be easily identified. As a standard spectrophotometer, measurement modes include photometrics, concentration, multi-wavelength, spectrum scanning, quantitation, and kinetics.

Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat